Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615932 MIM: 606964 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCTD20 - potassium channel tetramerization domain containing 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STK38 - serine/threonine kinase 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305102.1 | Intron | NP_001292031.1 | ||||
NM_007271.3 | Intron | NP_009202.1 | ||||
XM_006714988.3 | Intron | XP_006715051.1 | ||||
XM_006714989.3 | Intron | XP_006715052.1 | ||||
XM_017010226.1 | Intron | XP_016865715.1 |
Set Membership: |
HapMap |