Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGAGTGAATATCTTACCCTTGC[C/T]ATCCTGACACTCACTGGGGCCCAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602982 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNN1 - potassium calcium-activated channel subfamily N member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002248.4 | Intron | NP_002239.2 | ||||
XM_005259905.1 | Intron | XP_005259962.1 | ||||
XM_005259906.1 | Intron | XP_005259963.1 | ||||
XM_005259907.2 | Intron | XP_005259964.1 | ||||
XM_011528004.1 | Intron | XP_011526306.1 | ||||
XM_011528005.2 | Intron | XP_011526307.1 |
Set Membership: |
HapMap |