Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607703 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NUP210 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NUP210 - nucleoporin 210 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024923.3 | 6639 | UTR 3 | NP_079199.2 | |||
XM_005264994.3 | 6639 | Intron | XP_005265051.1 | |||
XM_011533538.2 | 6639 | Intron | XP_011531840.1 | |||
XM_017006018.1 | 6639 | UTR 3 | XP_016861507.1 | |||
XM_017006019.1 | 6639 | UTR 3 | XP_016861508.1 | |||
XM_017006020.1 | 6639 | UTR 3 | XP_016861509.1 | |||
XM_017006021.1 | 6639 | Intron | XP_016861510.1 |
Set Membership: |
HapMap |