Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAGCCAGCACAGGACAATGGAGC[G/T]GAGTGACTGACGCCTGATGATAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609290 MIM: 610346 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AK3 - adenylate kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199852.1 | Intron | NP_001186781.1 | ||||
NM_001199853.1 | Intron | NP_001186782.1 | ||||
NM_001199855.1 | Intron | NP_001186784.1 | ||||
NM_001199856.1 | Intron | NP_001186785.1 | ||||
NM_016282.3 | Intron | NP_057366.2 |
CDC37L1 - cell division cycle 37 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |