Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCTCCCGGGCGCAACTTCAGCTC[C/G]GTGCAGGGCTGCAGAGAGGTCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105379030 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105379030 - uncharacterized LOC105379030 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM171 - transmembrane protein 171 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161342.2 | 460 | Missense Mutation | CGT,GGT | R,G 86 | NP_001154814.1 | |
NM_173490.7 | 460 | Missense Mutation | CGT,GGT | R,G 86 | NP_775761.4 | |
XM_011543156.1 | 460 | Missense Mutation | CGT,GGT | R,G 86 | XP_011541458.1 |