Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTATACAGAGATTTGTCACACAT[A/T]GACATCTACATTTTCATGTATAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929574 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101929574 - uncharacterized LOC101929574 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC102723703 - uncharacterized LOC102723703 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH2D4B - SH2 domain containing 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145719.1 | 2190 | Intron | NP_001139191.1 | |||
NM_207372.2 | 2190 | Intron | NP_997255.2 | |||
XM_017016236.1 | 2190 | UTR 5 | XP_016871725.1 |
Set Membership: |
HapMap |