Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGACCTTCTGATGTTGGAGAAGG[C/T]GTGAGATCTGGGTATAGGTTCTCCC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FLJ26850 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FLJ26850 - FLJ26850 protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100287477 - zinc finger protein 420-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011527593.2 | 1466 | Missense Mutation | CAC,CGC | H,R 489 | XP_011525895.1 |
Set Membership: |
HapMap |