Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCTTTTGTTGCTTGTGCTTTTC[A/G]TTTCTTAGCTAAGAAAGAAGGGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611259 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDKAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDKAL1 - CDK5 regulatory subunit associated protein 1 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017774.3 | Intron | NP_060244.2 | ||||
XM_011514718.1 | Intron | XP_011513020.1 | ||||
XM_011514719.2 | Intron | XP_011513021.1 | ||||
XM_017010986.1 | Intron | XP_016866475.1 | ||||
XM_017010987.1 | Intron | XP_016866476.1 |