Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAGCACAGGCTTACTGCATGCCT[A/G]CTGTGTGTCACTTGCTGAGCTAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610112 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CMIP - c-Maf inducing protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030629.2 | Intron | NP_085132.1 | ||||
NM_198390.2 | Intron | NP_938204.2 | ||||
XM_005256179.4 | Intron | XP_005256236.4 | ||||
XM_005256181.2 | Intron | XP_005256238.1 | ||||
XM_005256182.1 | Intron | XP_005256239.1 | ||||
XM_011523352.1 | Intron | XP_011521654.1 | ||||
XM_011523353.1 | Intron | XP_011521655.1 | ||||
XM_017023733.1 | Intron | XP_016879222.1 |