Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCAAACCCTATTTAGTAATGACT[A/G]TTGGCTGTCTCAGTGATTTGATAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
40 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 182310 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP1A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
ATP1A1 - ATPase Na+/K+ transporting subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000701.7 | 444 | Intron | NP_000692.2 | |||
NM_001160233.1 | 444 | Intron | NP_001153705.1 | |||
NM_001160234.1 | 444 | Intron | NP_001153706.1 | |||
XM_017001360.1 | 444 | UTR 5 | XP_016856849.1 | |||
XM_017001361.1 | 444 | UTR 5 | XP_016856850.1 |
Set Membership: |
HapMap Validated |