Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAAGTTGTCCAAGATGTTATGGA[C/G]AAGTGAGGGAGAGGAGAGGAAGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606273 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EIF2B3 - eukaryotic translation initiation factor 2B subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166588.2 | Intron | NP_001160060.1 | ||||
NM_001261418.1 | Intron | NP_001248347.1 | ||||
NM_020365.4 | Intron | NP_065098.1 | ||||
XM_017002745.1 | Intron | XP_016858234.1 | ||||
XM_017002746.1 | Intron | XP_016858235.1 | ||||
XM_017002747.1 | Intron | XP_016858236.1 |
Set Membership: |
HapMap |