Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606273 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF2B3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF2B3 - eukaryotic translation initiation factor 2B subunit gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166588.2 | Intron | NP_001160060.1 | ||||
NM_001261418.1 | Intron | NP_001248347.1 | ||||
NM_020365.4 | Intron | NP_065098.1 | ||||
XM_017002745.1 | Intron | XP_016858234.1 | ||||
XM_017002746.1 | Intron | XP_016858235.1 | ||||
XM_017002747.1 | Intron | XP_016858236.1 |