Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGTCTCTTTTTAAAAAAATAGCA[C/T]TTATCACCTAATACACACTACACAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613187 MIM: 606161 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR103A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR103A1 - microRNA 103a-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR103B1 - microRNA 103b-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PANK3 - pantothenate kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024594.3 | Intron | NP_078870.1 |
Set Membership: |
HapMap |