Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605252 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLI PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
POLI - DNA polymerase iota | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007195.2 | Intron | NP_009126.2 | ||||
XM_005258188.3 | Intron | XP_005258245.1 | ||||
XM_005258192.4 | Intron | XP_005258249.1 | ||||
XM_006722378.2 | Intron | XP_006722441.1 | ||||
XM_011525796.2 | Intron | XP_011524098.1 | ||||
XM_011525797.1 | Intron | XP_011524099.1 |
Set Membership: |
HapMap |