Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
80 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605554 MIM: 600684 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD244 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CD244 - CD244 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166663.1 | 1995 | UTR 3 | NP_001160135.1 | |||
NM_001166664.1 | 1995 | UTR 3 | NP_001160136.1 | |||
NM_016382.3 | 1995 | UTR 3 | NP_057466.1 | |||
XM_011509621.2 | 1995 | Intron | XP_011507923.1 | |||
XM_011509622.2 | 1995 | Intron | XP_011507924.1 | |||
XM_011509623.2 | 1995 | UTR 3 | XP_011507925.1 |
LY9 - lymphocyte antigen 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP Validated |