Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATCGGTTCAGGAGTTTAGATCCA[C/T]TCACGGATACTGACCTGTCACCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602730 MIM: 604049 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACVR2B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ACVR2B - activin A receptor type 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001106.3 | 1547 | Intron | NP_001097.2 | |||
XM_005265583.3 | 1547 | UTR 5 | XP_005265640.1 | |||
XM_017007514.1 | 1547 | UTR 5 | XP_016863003.1 | |||
XM_017007515.1 | 1547 | Intron | XP_016863004.1 | |||
XM_017007516.1 | 1547 | Intron | XP_016863005.1 |
ACVR2B-AS1 - ACVR2B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XYLB - xylulokinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |