Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGCATCTTTCCCCTCTCTTGTC[C/T]CTGGGTGCTGAAGGATGTCACACTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608214 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAMD1B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GRAMD1B - GRAM domain containing 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCN3B - sodium voltage-gated channel beta subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040151.1 | 3200 | UTR 3 | NP_001035241.1 | |||
NM_018400.3 | 3200 | UTR 3 | NP_060870.1 | |||
XM_011542897.1 | 3200 | Intron | XP_011541199.1 |
Set Membership: |
HapMap |