Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCTCCTGGTGGGCGGCTATACC[C/T]CCCCAGGTCCAGGTTTCATTCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 107710 MIM: 107741 MIM: 608061 | ||||||||||||||||||||
Literature Links: |
APOC1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOC1 - apolipoprotein C1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOE - apolipoprotein E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000041.3 | Intron | NP_000032.1 | ||||
NM_001302688.1 | Intron | NP_001289617.1 | ||||
NM_001302689.1 | Intron | NP_001289618.1 | ||||
NM_001302690.1 | Intron | NP_001289619.1 | ||||
NM_001302691.1 | Intron | NP_001289620.1 |
TOMM40 - translocase of outer mitochondrial membrane 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |