Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAGGGTCACCAAAAAACTCTCT[C/G]AAGAGGGCCTGATTTAAGTATATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603521 MIM: 607918 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNRPA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNRPA1 - small nuclear ribonucleoprotein polypeptide A' | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VIMP - VCP interacting membrane selenoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018445.5 | Intron | NP_060915.2 | ||||
NM_203472.2 | Intron | NP_982298.2 |