Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCTACAGGAGGCTGAGGGTCTGGC[A/G]GCGGAAGGGCAGGAGCCTCGGACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609722 MIM: 610795 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDLIM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDLIM2 - PDZ and LIM domain 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021630.5 | Intron | NP_067643.3 | ||||
NM_176871.3 | Intron | NP_789847.1 | ||||
NM_198042.3 | Intron | NP_932159.1 |
SORBS3 - sorbin and SH3 domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |