Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612993 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CMSS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CMSS1 - cms1 ribosomal small subunit homolog (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167924.1 | Intron | NP_001161396.1 | ||||
NM_032359.3 | Intron | NP_115735.2 |
FILIP1L - filamin A interacting protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042459.2 | Intron | NP_001035924.1 | ||||
NM_001282793.1 | Intron | NP_001269722.1 | ||||
NM_001282794.1 | Intron | NP_001269723.1 | ||||
NM_014890.3 | Intron | NP_055705.2 | ||||
NM_182909.3 | Intron | NP_878913.2 | ||||
XM_006713486.2 | Intron | XP_006713549.1 | ||||
XM_011512371.2 | Intron | XP_011510673.1 |
Set Membership: |
HapMap |