Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608116 MIM: 615340 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HHATL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HHATL - hedgehog acyltransferase-like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020707.3 | 1423 | Missense Mutation | ATG,GTG | M,V 403 | NP_065758.3 | |
XM_006713274.2 | 1423 | Missense Mutation | ATG,GTG | M,V 403 | XP_006713337.1 | |
XM_006713275.2 | 1423 | Missense Mutation | ATG,GTG | M,V 338 | XP_006713338.1 | |
XM_011533969.2 | 1423 | Missense Mutation | ATG,GTG | M,V 426 | XP_011532271.1 | |
XM_011533970.2 | 1423 | Missense Mutation | ATG,GTG | M,V 361 | XP_011532272.1 | |
XM_017006935.1 | 1423 | Missense Mutation | ATG,GTG | M,V 426 | XP_016862424.1 | |
XM_017006936.1 | 1423 | Missense Mutation | ATG,GTG | M,V 403 | XP_016862425.1 | |
XM_017006937.1 | 1423 | Missense Mutation | ATG,GTG | M,V 338 | XP_016862426.1 | |
XM_017006938.1 | 1423 | Missense Mutation | ATG,GTG | M,V 338 | XP_016862427.1 |
KLHL40 - kelch like family member 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |