Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614829 MIM: 612535 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4orf26 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C4orf26 - chromosome 4 open reading frame 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206981.1 | 749 | Intron | NP_001193910.1 | |||
NM_001257072.1 | 749 | Intron | NP_001244001.1 | |||
NM_178497.3 | 749 | Intron | NP_848592.2 | |||
XM_011531668.2 | 749 | UTR 5 | XP_011529970.1 |
THAP6 - THAP domain containing 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |