Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602776 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
REV3L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
REV3L - REV3 like, DNA directed polymerase zeta catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286431.1 | Intron | NP_001273360.1 | ||||
NM_001286432.1 | Intron | NP_001273361.1 | ||||
NM_002912.4 | Intron | NP_002903.3 | ||||
XM_011536028.1 | Intron | XP_011534330.1 | ||||
XM_011536029.2 | Intron | XP_011534331.1 | ||||
XM_011536030.2 | Intron | XP_011534332.1 | ||||
XM_011536031.2 | Intron | XP_011534333.1 | ||||
XM_011536032.2 | Intron | XP_011534334.1 | ||||
XM_011536036.2 | Intron | XP_011534338.1 | ||||
XM_017011152.1 | Intron | XP_016866641.1 | ||||
XM_017011153.1 | Intron | XP_016866642.1 | ||||
XM_017011154.1 | Intron | XP_016866643.1 | ||||
XM_017011155.1 | Intron | XP_016866644.1 |
Set Membership: |
HapMap Validated |