Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608216 MIM: 194531 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COMMD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COMMD5 - COMM domain containing 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF7 - zinc finger protein 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282795.1 | 327 | Intron | NP_001269724.1 | |||
NM_001282796.1 | 327 | Intron | NP_001269725.1 | |||
NM_001282797.1 | 327 | Intron | NP_001269726.1 | |||
NM_003416.3 | 327 | Intron | NP_003407.1 | |||
XM_006716654.2 | 327 | Intron | XP_006716717.1 | |||
XM_006716656.3 | 327 | Intron | XP_006716719.1 | |||
XM_011517291.2 | 327 | Intron | XP_011515593.1 | |||
XM_011517292.2 | 327 | Intron | XP_011515594.1 | |||
XM_011517293.2 | 327 | Intron | XP_011515595.1 | |||
XM_011517294.2 | 327 | Intron | XP_011515596.1 | |||
XM_011517295.2 | 327 | Intron | XP_011515597.1 | |||
XM_011517296.2 | 327 | Intron | XP_011515598.1 | |||
XM_011517297.2 | 327 | Intron | XP_011515599.1 | |||
XM_017013813.1 | 327 | Intron | XP_016869302.1 | |||
XM_017013814.1 | 327 | UTR 5 | XP_016869303.1 | |||
XM_017013815.1 | 327 | UTR 5 | XP_016869304.1 | |||
XM_017013816.1 | 327 | Intron | XP_016869305.1 | |||
XM_017013817.1 | 327 | Intron | XP_016869306.1 |