Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
58 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176265 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNC4 - potassium voltage-gated channel subfamily C member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039574.2 | Intron | NP_001034663.1 | ||||
NM_004978.4 | Intron | NP_004969.2 | ||||
XM_006710625.3 | Intron | XP_006710688.1 | ||||
XM_006710627.3 | Intron | XP_006710690.1 | ||||
XM_011541401.2 | Intron | XP_011539703.1 | ||||
XM_011541402.1 | Intron | XP_011539704.1 | ||||
XM_011541403.1 | Intron | XP_011539705.1 | ||||
XM_011541404.1 | Intron | XP_011539706.1 |
KCNC4-AS1 - KCNC4 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap JSNP |