Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610872 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF19B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF19B - ring finger protein 19B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127361.2 | Intron | NP_001120833.1 | ||||
NM_001300826.1 | Intron | NP_001287755.1 | ||||
NM_153341.3 | Intron | NP_699172.2 | ||||
XM_006710356.2 | Intron | XP_006710419.1 | ||||
XM_006710357.3 | Intron | XP_006710420.1 | ||||
XM_006710358.3 | Intron | XP_006710421.1 |
Set Membership: |
HapMap |