Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611505 MIM: 603479 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CENPP PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CENPP - centromere protein P | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012267.2 | Intron | NP_001012267.1 | ||||
NM_001286969.1 | Intron | NP_001273898.1 | ||||
NM_001286971.1 | Intron | NP_001273900.1 | ||||
XM_011518685.2 | Intron | XP_011516987.1 | ||||
XM_011518689.1 | Intron | XP_011516991.1 | ||||
XM_017014715.1 | Intron | XP_016870204.1 |
ECM2 - extracellular matrix protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197295.1 | Intron | NP_001184224.1 | ||||
NM_001197296.1 | Intron | NP_001184225.1 | ||||
NM_001393.3 | Intron | NP_001384.1 | ||||
XM_017014376.1 | Intron | XP_016869865.1 | ||||
XM_017014377.1 | Intron | XP_016869866.1 |