Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608003 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIPK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIPK1 - homeodomain interacting protein kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152696.3 | Intron | NP_689909.2 | ||||
NM_181358.2 | Intron | NP_852003.1 | ||||
NM_198268.2 | Intron | NP_938009.1 | ||||
NM_198269.2 | Intron | NP_938010.1 | ||||
XM_005270609.3 | Intron | XP_005270666.1 | ||||
XM_005270610.4 | Intron | XP_005270667.1 | ||||
XM_005270611.4 | Intron | XP_005270668.1 | ||||
XM_005270612.4 | Intron | XP_005270669.1 | ||||
XM_005270613.4 | Intron | XP_005270670.1 | ||||
XM_006710443.3 | Intron | XP_006710506.1 | ||||
XM_011540977.2 | Intron | XP_011539279.1 | ||||
XM_017000605.1 | Intron | XP_016856094.1 | ||||
XM_017000606.1 | Intron | XP_016856095.1 |
HIPK1-AS1 - HIPK1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |