Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611089 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTMR14 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MTMR14 - myotubularin related protein 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077525.2 | Intron | NP_001070993.1 | ||||
NM_001077526.2 | Intron | NP_001070994.1 | ||||
NM_022485.4 | Intron | NP_071930.2 | ||||
XM_005265400.1 | Intron | XP_005265457.1 | ||||
XM_011534012.1 | Intron | XP_011532314.1 | ||||
XM_011534013.1 | Intron | XP_011532315.1 | ||||
XM_017007034.1 | Intron | XP_016862523.1 | ||||
XM_017007035.1 | Intron | XP_016862524.1 | ||||
XM_017007036.1 | Intron | XP_016862525.1 | ||||
XM_017007037.1 | Intron | XP_016862526.1 | ||||
XM_017007038.1 | Intron | XP_016862527.1 | ||||
XM_017007039.1 | Intron | XP_016862528.1 | ||||
XM_017007040.1 | Intron | XP_016862529.1 | ||||
XM_017007041.1 | Intron | XP_016862530.1 | ||||
XM_017007042.1 | Intron | XP_016862531.1 | ||||
XM_017007043.1 | Intron | XP_016862532.1 | ||||
XM_017007044.1 | Intron | XP_016862533.1 | ||||
XM_017007045.1 | Intron | XP_016862534.1 |
Set Membership: |
HapMap |