Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 125264 MIM: 613081 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DEK PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
DEK - DEK proto-oncogene | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134709.1 | 2698 | UTR 3 | NP_001128181.1 | |||
NM_003472.3 | 2698 | UTR 3 | NP_003463.1 | |||
XM_017011273.1 | 2698 | Intron | XP_016866762.1 |
KDM1B - lysine demethylase 1B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153042.3 | 2698 | Intron | NP_694587.3 | |||
XM_005248925.1 | 2698 | Intron | XP_005248982.1 | |||
XM_005248926.1 | 2698 | Intron | XP_005248983.1 | |||
XM_005248928.1 | 2698 | Intron | XP_005248985.1 | |||
XM_011514385.1 | 2698 | Intron | XP_011512687.1 | |||
XM_011514386.1 | 2698 | Intron | XP_011512688.1 | |||
XM_017010440.1 | 2698 | Intron | XP_016865929.1 | |||
XM_017010441.1 | 2698 | Intron | XP_016865930.1 | |||
XM_017010442.1 | 2698 | Intron | XP_016865931.1 | |||
XM_017010443.1 | 2698 | Intron | XP_016865932.1 | |||
XM_017010445.1 | 2698 | Intron | XP_016865934.1 | |||
XM_017010446.1 | 2698 | Intron | XP_016865935.1 |
Set Membership: |
HapMap |