Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612328 MIM: 609641 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC73 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC73 - coiled-coil domain containing 73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008391.3 | 1310 | Silent Mutation | AAA,AAG | K,K 1064 | NP_001008392.2 | |
XM_011520139.2 | 1310 | Silent Mutation | AAA,AAG | K,K 978 | XP_011518441.1 |
EIF3M - eukaryotic translation initiation factor 3 subunit M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307929.1 | 1310 | UTR 3 | NP_001294858.1 | |||
NM_006360.5 | 1310 | UTR 3 | NP_006351.2 | |||
XM_011519840.2 | 1310 | Intron | XP_011518142.1 |