Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTATTGGGTAATTTTAAAAATAG[C/T]GTGTTAAATAACTTTTTCATTTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612328 MIM: 609641 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC73 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC73 - coiled-coil domain containing 73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008391.3 | Intron | NP_001008392.2 | ||||
XM_011520139.2 | Intron | XP_011518441.1 |
EIF3M - eukaryotic translation initiation factor 3 subunit M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001307929.1 | Intron | NP_001294858.1 | ||||
NM_006360.5 | Intron | NP_006351.2 | ||||
XM_011519840.2 | Intron | XP_011518142.1 |