Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602221 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZMYM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ZMYM2 - zinc finger MYM-type containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001190964.2 | Intron | NP_001177893.1 | ||||
NM_001190965.2 | Intron | NP_001177894.1 | ||||
NM_003453.4 | Intron | NP_003444.1 | ||||
NM_197968.3 | Intron | NP_932072.1 | ||||
XM_005266520.3 | Intron | XP_005266577.1 | ||||
XM_011535216.2 | Intron | XP_011533518.1 | ||||
XM_011535222.1 | Intron | XP_011533524.1 | ||||
XM_011535223.2 | Intron | XP_011533525.1 | ||||
XM_011535224.2 | Intron | XP_011533526.1 | ||||
XM_017020728.1 | Intron | XP_016876217.1 | ||||
XM_017020729.1 | Intron | XP_016876218.1 | ||||
XM_017020731.1 | Intron | XP_016876220.1 | ||||
XM_017020732.1 | Intron | XP_016876221.1 | ||||
XM_017020733.1 | Intron | XP_016876222.1 | ||||
XM_017020734.1 | Intron | XP_016876223.1 | ||||
XM_017020735.1 | Intron | XP_016876224.1 | ||||
XM_017020736.1 | Intron | XP_016876225.1 | ||||
XM_017020737.1 | Intron | XP_016876226.1 | ||||
XM_017020738.1 | Intron | XP_016876227.1 | ||||
XM_017020739.1 | Intron | XP_016876228.1 |
Set Membership: |
HapMap |