Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613927 MIM: 138470 MIM: 154040 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
C2 - complement component 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C2-AS1 - C2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CFB - complement factor B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001710.5 | 728 | Silent Mutation | CGA,CGG | R,R 150 | NP_001701.2 |
MIR1236 - microRNA 1236 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NELFE - negative elongation factor complex member E | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |