Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610464 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR156 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPR156 - G protein-coupled receptor 156 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168271.1 | 4123 | UTR 3 | NP_001161743.1 | |||
NM_153002.2 | 4123 | UTR 3 | NP_694547.2 | |||
XM_005247165.2 | 4123 | UTR 3 | XP_005247222.1 | |||
XM_011512484.2 | 4123 | UTR 3 | XP_011510786.1 | |||
XM_011512487.1 | 4123 | UTR 3 | XP_011510789.1 | |||
XM_011512488.2 | 4123 | UTR 3 | XP_011510790.1 | |||
XM_017005795.1 | 4123 | UTR 3 | XP_016861284.1 | |||
XM_017005796.1 | 4123 | UTR 3 | XP_016861285.1 | |||
XM_017005797.1 | 4123 | UTR 3 | XP_016861286.1 |
Set Membership: |
HapMap |