Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGAATGGAAAAGTTGGCTGGCAT[C/T]GGATTGTGGAGGCCTTGAAAGCCTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 600552 | |||||||||||||||||||||||
Literature Links: |
XCR1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
XCR1 - X-C motif chemokine receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024644.1 | 2542 | Intron | NP_001019815.1 | |||
NM_005283.2 | 2542 | Intron | NP_005274.1 | |||
XM_017006196.1 | 2542 | UTR 3 | XP_016861685.1 | |||
XM_017006197.1 | 2542 | UTR 3 | XP_016861686.1 | |||
XM_017006198.1 | 2542 | UTR 3 | XP_016861687.1 | |||
XM_017006199.1 | 2542 | UTR 3 | XP_016861688.1 | |||
XM_017006200.1 | 2542 | UTR 3 | XP_016861689.1 | |||
XM_017006201.1 | 2542 | UTR 3 | XP_016861690.1 |