Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAATGGCATCTTGAACAGTGCAA[C/T]GTAGGATATATAATTGCGGGTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASSF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RASSF8 - Ras association domain family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164746.1 | Intron | NP_001158218.1 | ||||
NM_001164747.1 | Intron | NP_001158219.1 | ||||
NM_001164748.1 | Intron | NP_001158220.1 | ||||
NM_007211.4 | Intron | NP_009142.2 | ||||
XM_006719027.2 | Intron | XP_006719090.1 | ||||
XM_006719028.1 | Intron | XP_006719091.1 | ||||
XM_017018746.1 | Intron | XP_016874235.1 |
RASSF8-AS1 - RASSF8 atnisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |