Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTGGCTCTTGTAGGCTTCTATAC[A/G]TGGTGACTTACGTCAGTTTTTGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608231 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RASSF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RASSF8 - Ras association domain family member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164746.1 | Intron | NP_001158218.1 | ||||
NM_001164747.1 | Intron | NP_001158219.1 | ||||
NM_001164748.1 | Intron | NP_001158220.1 | ||||
NM_007211.4 | Intron | NP_009142.2 | ||||
XM_006719027.2 | Intron | XP_006719090.1 | ||||
XM_006719028.1 | Intron | XP_006719091.1 | ||||
XM_017018746.1 | Intron | XP_016874235.1 |
RASSF8-AS1 - RASSF8 atnisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |