Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602709 MIM: 607608 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APBB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APBB1 - amyloid beta precursor protein binding family B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164.4 | Intron | NP_001155.1 | ||||
NM_001257319.2 | Intron | NP_001244248.1 | ||||
NM_001257320.2 | Intron | NP_001244249.1 | ||||
NM_001257321.2 | Intron | NP_001244250.1 | ||||
NM_001257323.2 | Intron | NP_001244252.1 | ||||
NM_001257325.2 | Intron | NP_001244254.1 | ||||
NM_001257326.2 | Intron | NP_001244255.1 | ||||
NM_145689.2 | Intron | NP_663722.1 | ||||
XM_006718209.3 | Intron | XP_006718272.2 | ||||
XM_006718210.3 | Intron | XP_006718273.2 | ||||
XM_006718211.1 | Intron | XP_006718274.1 | ||||
XM_006718212.3 | Intron | XP_006718275.2 | ||||
XM_006718213.1 | Intron | XP_006718276.1 | ||||
XM_006718214.3 | Intron | XP_006718277.2 | ||||
XM_006718215.3 | Intron | XP_006718278.3 | ||||
XM_011520037.2 | Intron | XP_011518339.1 | ||||
XM_011520038.2 | Intron | XP_011518340.1 | ||||
XM_011520040.2 | Intron | XP_011518342.1 | ||||
XM_011520042.1 | Intron | XP_011518344.1 | ||||
XM_017017639.1 | Intron | XP_016873128.1 | ||||
XM_017017640.1 | Intron | XP_016873129.1 | ||||
XM_017017641.1 | Intron | XP_016873130.1 | ||||
XM_017017642.1 | Intron | XP_016873131.1 |
SMPD1 - sphingomyelin phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |