Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608858 MIM: 605181 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGFR1OP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FGFR1OP2 - FGFR1 oncogene partner 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TM7SF3 - transmembrane 7 superfamily member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016551.2 | 4084 | UTR 3 | NP_057635.1 | |||
XM_005253391.3 | 4084 | UTR 3 | XP_005253448.1 | |||
XM_011520690.2 | 4084 | UTR 3 | XP_011518992.1 | |||
XM_011520691.1 | 4084 | Intron | XP_011518993.1 | |||
XM_017019463.1 | 4084 | UTR 3 | XP_016874952.1 | |||
XM_017019464.1 | 4084 | UTR 3 | XP_016874953.1 | |||
XM_017019465.1 | 4084 | UTR 3 | XP_016874954.1 | |||
XM_017019466.1 | 4084 | UTR 3 | XP_016874955.1 |