Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGATAACCACCATTCACAGGAAGA[A/G]TATAGAACCCAGGAAGTTTTCCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLRX3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLRX3 - glutaredoxin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199868.1 | Intron | NP_001186797.1 | ||||
NM_001321980.1 | Intron | NP_001308909.1 | ||||
NM_006541.4 | Intron | NP_006532.2 | ||||
XM_017015474.1 | Intron | XP_016870963.1 |