Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601819 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BPTF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BPTF - bromodomain PHD finger transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004459.6 | Intron | NP_004450.3 | ||||
NM_182641.3 | Intron | NP_872579.2 | ||||
XM_005257150.3 | Intron | XP_005257207.1 | ||||
XM_005257151.3 | Intron | XP_005257208.1 | ||||
XM_005257152.2 | Intron | XP_005257209.1 | ||||
XM_005257153.3 | Intron | XP_005257210.1 | ||||
XM_005257154.3 | Intron | XP_005257211.1 | ||||
XM_005257155.3 | Intron | XP_005257212.1 | ||||
XM_005257156.3 | Intron | XP_005257213.1 | ||||
XM_005257157.3 | Intron | XP_005257214.1 | ||||
XM_005257158.3 | Intron | XP_005257215.1 | ||||
XM_005257159.2 | Intron | XP_005257216.1 | ||||
XM_005257160.2 | Intron | XP_005257217.1 | ||||
XM_005257161.3 | Intron | XP_005257218.1 | ||||
XM_011524520.2 | Intron | XP_011522822.1 | ||||
XM_011524521.2 | Intron | XP_011522823.1 | ||||
XM_011524522.2 | Intron | XP_011522824.1 | ||||
XM_011524523.2 | Intron | XP_011522825.1 | ||||
XM_011524524.2 | Intron | XP_011522826.1 | ||||
XM_011524525.2 | Intron | XP_011522827.1 | ||||
XM_011524526.2 | Intron | XP_011522828.1 | ||||
XM_017024353.1 | Intron | XP_016879842.1 | ||||
XM_017024354.1 | Intron | XP_016879843.1 |