Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAACAGATTCTGATTTATAATAC[A/G]AGTGTTGGAGTGAACGTCAAAACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614295 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BICC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BICC1 - BicC family RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080512.2 | Intron | NP_001073981.1 | ||||
XM_005270169.4 | Intron | XP_005270226.1 | ||||
XM_011540185.2 | Intron | XP_011538487.1 | ||||
XM_011540188.2 | Intron | XP_011538490.2 | ||||
XM_011540189.2 | Intron | XP_011538491.1 | ||||
XM_011540190.2 | Intron | XP_011538492.1 | ||||
XM_011540191.2 | Intron | XP_011538493.1 | ||||
XM_017016677.1 | Intron | XP_016872166.1 | ||||
XM_017016678.1 | Intron | XP_016872167.1 |
Set Membership: |
Validated |