Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCTTTATGCATAACTTACTTTGA[C/T]TTTATCAGTCAATGCATCGCAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614295 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BICC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BICC1 - BicC family RNA binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080512.2 | Intron | NP_001073981.1 | ||||
XM_005270169.4 | Intron | XP_005270226.1 | ||||
XM_011540185.2 | Intron | XP_011538487.1 | ||||
XM_011540188.2 | Intron | XP_011538490.2 | ||||
XM_011540189.2 | Intron | XP_011538491.1 | ||||
XM_011540190.2 | Intron | XP_011538492.1 | ||||
XM_011540191.2 | Intron | XP_011538493.1 | ||||
XM_017016677.1 | Intron | XP_016872166.1 | ||||
XM_017016678.1 | Intron | XP_016872167.1 |