Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGATAACATTCAGTAACTTTTTAA[C/T]AACAACTTGAATACATTACATTTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616031 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC141 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC141 - coiled-coil domain containing 141 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001316745.1 | 6488 | Intron | NP_001303674.1 | |||
NM_173648.3 | 6488 | UTR 3 | NP_775919.3 | |||
XM_011510991.2 | 6488 | UTR 3 | XP_011509293.1 | |||
XM_011510992.2 | 6488 | Intron | XP_011509294.1 | |||
XM_011510993.2 | 6488 | Intron | XP_011509295.1 | |||
XM_017003867.1 | 6488 | Intron | XP_016859356.1 | |||
XM_017003868.1 | 6488 | Intron | XP_016859357.1 | |||
XM_017003869.1 | 6488 | Intron | XP_016859358.1 | |||
XM_017003870.1 | 6488 | Intron | XP_016859359.1 | |||
XM_017003871.1 | 6488 | Intron | XP_016859360.1 |