Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610701 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ENO4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ENO4 - enolase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242699.1 | Intron | NP_001229628.1 | ||||
XM_005269815.3 | Intron | XP_005269872.1 | ||||
XM_006717835.3 | Intron | XP_006717898.1 | ||||
XM_006717836.3 | Intron | XP_006717899.1 | ||||
XM_011539797.2 | Intron | XP_011538099.1 | ||||
XM_011539799.2 | Intron | XP_011538101.1 | ||||
XM_017016241.1 | Intron | XP_016871730.1 |
HSPA12A - heat shock protein family A (Hsp70) member 12A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025015.2 | Intron | NP_079291.2 | ||||
XM_005269672.3 | Intron | XP_005269729.1 | ||||
XM_005269673.4 | Intron | XP_005269730.1 | ||||
XM_011539579.2 | Intron | XP_011537881.1 | ||||
XM_011539580.2 | Intron | XP_011537882.1 | ||||
XM_017016032.1 | Intron | XP_016871521.1 |
Set Membership: |
HapMap Validated |