Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTCCCCACCTGGGATCTTTGTG[C/G]CTTTCTCCAAATCCTTCTGCCAGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606495 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARK4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MARK4 - microtubule affinity regulating kinase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199867.1 | Intron | NP_001186796.1 | ||||
NM_031417.3 | Intron | NP_113605.2 | ||||
XM_006723307.3 | Intron | XP_006723370.1 |
Set Membership: |
HapMap Validated |