Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 200350 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACACA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACACA - acetyl-CoA carboxylase alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198834.2 | Intron | NP_942131.1 | ||||
NM_198836.2 | Intron | NP_942133.1 | ||||
NM_198837.1 | Intron | NP_942134.1 | ||||
NM_198838.1 | Intron | NP_942135.1 | ||||
NM_198839.2 | Intron | NP_942136.1 | ||||
XM_005257267.4 | Intron | XP_005257324.1 | ||||
XM_006721853.1 | Intron | XP_006721916.1 | ||||
XM_011524701.1 | Intron | XP_011523003.1 | ||||
XM_011524703.1 | Intron | XP_011523005.1 | ||||
XM_011524704.2 | Intron | XP_011523006.1 | ||||
XM_017024553.1 | Intron | XP_016880042.1 | ||||
XM_017024554.1 | Intron | XP_016880043.1 | ||||
XM_017024555.1 | Intron | XP_016880044.1 |
Set Membership: |
HapMap |