Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTTATTGCATCATGAAGGCAC[A/G]GTAAGATCATTTGGCAAATTGTCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164757 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BRAF PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | |||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
BRAF - B-Raf proto-oncogene, serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004333.4 | 6338 | Intron | NP_004324.2 | |||
XM_005250045.2 | 6338 | UTR 3 | XP_005250102.1 | |||
XM_017012558.1 | 6338 | UTR 3 | XP_016868047.1 | |||
XM_017012559.1 | 6338 | UTR 3 | XP_016868048.1 |
Set Membership: |
HapMap Validated |